Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005789.hg.1 |
ENST00000300504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005789.hg.1 |
ENST00000535547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005789.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005789.hg.1 |
TC15000668.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007825.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155619.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155619.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155619.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155619.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155619.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155619.hg.1 |
ENST00000300504.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155619.hg.1 |
ENST00000535547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155619.hg.1 |
ENST00000562056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155619.hg.1 |
NM_001286434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155619.hg.1 |
NM_153356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155619.hg.1 |
TC1500007825.hg.1 |
Clariom D TC |