Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024589.hg.1 |
TC06001984.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024589.hg.1 |
ENST00000369063.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024589.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008240.hg.1 |
ENST00000553021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008240.hg.1 |
ENST00000553021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008240.hg.1 |
ENST00000553021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008240.hg.1 |
ENST00000553021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000493619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000493619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000498091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000498091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000498091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000539449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000539449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000539449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000539449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000539449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
ENST00000539449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187835.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187835.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187835.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187835.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187835.hg.1 |
TC0600012740.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200008240.hg.1 |
ENST00000553021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008240.hg.1 |
ENST00000553021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008240.hg.1 |
ENST00000553021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008240.hg.1 |
ENST00000553021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000369063.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000493619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000493619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000498091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000498091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000498091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000539449.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000539449.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000539449.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000539449.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000539449.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
ENST00000539449.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012740.hg.1 |
NM_032730.5 |
Refseq |