Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023855.hg.1 |
TC01001545.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023855.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023855.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
ENST00000440702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
ENST00000440702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
ENST00000440702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
ENST00000440702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100179003.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179003.hg.1 |
ENST00000440702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179003.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100179003.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179003.hg.1 |
ENST00000440702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179003.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100179003.hg.1 |
TC0100010761.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
ENST00000263733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
ENST00000440702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
ENST00000440702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
ENST00000440702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
ENST00000440702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010761.hg.1 |
NM_014864.3 |
Refseq |