Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020743.hg.1 |
TC09001634.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020743.hg.1 |
ENST00000372715.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020743.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000419989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000419989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000419989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000419989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000451652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000451652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000451652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000451652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000451652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000451652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000473486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000473486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000473486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000473486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000480613.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000480613.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000480613.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000480613.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000480613.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000483181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000483181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000483181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000483181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000483181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
ENST00000483181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000419989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000419989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000419989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000419989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000451652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000451652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000451652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000451652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000451652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000451652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000473486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000473486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000473486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000473486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000480613.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000480613.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000480613.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000480613.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000480613.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000483181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000483181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000483181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000483181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000483181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
ENST00000483181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011650.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900179680.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179680.hg.1 |
ENST00000419989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179680.hg.1 |
ENST00000451652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179680.hg.1 |
ENST00000473486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179680.hg.1 |
ENST00000480613.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179680.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900179680.hg.1 |
ENST00000372715.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179680.hg.1 |
ENST00000419989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179680.hg.1 |
ENST00000451652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179680.hg.1 |
ENST00000473486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179680.hg.1 |
ENST00000480613.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179680.hg.1 |
NM_052844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900179680.hg.1 |
TC0900011650.hg.1 |
Clariom D TC |