Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026834.hg.1 |
TC03001473.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026834.hg.1 |
ENST00000335754.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026834.hg.1 |
ENST00000482349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026834.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000488802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000488802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000488802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000488802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000488802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000495993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000495993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000495993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000495993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000498740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000498740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000498740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000498740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000498740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000498740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000498740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
ENST00000498740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184057.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184057.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184057.hg.1 |
ENST00000498740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184057.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184057.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184057.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184057.hg.1 |
ENST00000498740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184057.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184057.hg.1 |
TC0300011222.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000335754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000482349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000486794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000488802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000488802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000488802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000488802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000488802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000495993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000495993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000495993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000495993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000498740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000498740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000498740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000498740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000498740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000498740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000498740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
ENST00000498740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011222.hg.1 |
NM_022899.4 |
Refseq |