Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000447516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000447516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000447516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000447516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005724.hg.1 |
TC07000425.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005724.hg.1 |
ENST00000333538.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005724.hg.1 |
ENST00000498380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005724.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700154684.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700154684.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700154684.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700154684.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700154684.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700154684.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700154684.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700154684.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700154684.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700154684.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700154684.hg.1 |
TC0700007925.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000333538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000447516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000447516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000447516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000447516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000467723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000498380.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
ENST00000618959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007925.hg.1 |
NM_022479.2 |
Refseq |