Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003175.hg.1 |
TC6_cox_hap2000171.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003175.hg.1 |
NM_006295.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001233.hg.1 |
TC6_apd_hap1000085.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001233.hg.1 |
NM_006295.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003082.hg.1 |
TC6_dbb_hap3000160.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003082.hg.1 |
NM_006295.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002825.hg.1 |
TC6_ssto_hap7000143.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002825.hg.1 |
NM_006295.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017870.hg.1 |
TC06001543.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017870.hg.1 |
ENST00000375663.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017870.hg.1 |
ENST00000482996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017870.hg.1 |
NM_006295.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002842.hg.1 |
TC6_mcf_hap5000148.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002842.hg.1 |
NM_006295.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600177649.hg.1 |
ENST00000375663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177649.hg.1 |
ENST00000428445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177649.hg.1 |
ENST00000459667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177649.hg.1 |
ENST00000461874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177649.hg.1 |
ENST00000479051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177649.hg.1 |
ENST00000482996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177649.hg.1 |
ENST00000375663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177649.hg.1 |
ENST00000428445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177649.hg.1 |
ENST00000459667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177649.hg.1 |
ENST00000461874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177649.hg.1 |
ENST00000479051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177649.hg.1 |
ENST00000482996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177649.hg.1 |
TC0600011455.hg.1 |
Clariom D TC |