Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027291.hg.1 |
ENST00000302528.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027291.hg.1 |
ENST00000358808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027291.hg.1 |
ENST00000536954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027291.hg.1 |
ENST00000540338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027291.hg.1 |
ENST00000542885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027291.hg.1 |
ENST00000545889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027291.hg.1 |
NM_001247997.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027291.hg.1 |
NM_002956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027291.hg.1 |
TC12002073.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200195715.hg.1 |
ENST00000302528.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195715.hg.1 |
ENST00000358808.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195715.hg.1 |
ENST00000540304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195715.hg.1 |
ENST00000540338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195715.hg.1 |
ENST00000545889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195715.hg.1 |
ENST00000620786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195715.hg.1 |
NM_001247997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195715.hg.1 |
NM_002956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195715.hg.1 |
ENST00000302528.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195715.hg.1 |
ENST00000358808.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195715.hg.1 |
ENST00000540304.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195715.hg.1 |
ENST00000540338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195715.hg.1 |
ENST00000545889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195715.hg.1 |
ENST00000620786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195715.hg.1 |
NM_001247997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195715.hg.1 |
NM_002956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195715.hg.1 |
TC1200012234.hg.1 |
Clariom D TC |