Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD21L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136566.2 | NP_001130038.2 | 9 - 10 | 1177 | 75 | |
XM_006723605.3 | XP_006723668.1 | 9 - 10 | 1214 | 75 | ||
GenBank mRNA | BC157891.1 | - | 9 - 10 | 1177 | 8862712 | 75 |
BC171911.1 | - | 9 - 10 | 1177 | 9054565 | 75 | |
HQ603827.1 | - | 8 - 9 | 1092 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |