Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013273.hg.1 |
ENST00000339475.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013273.hg.1 |
ENST00000408990.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013273.hg.1 |
ENST00000555006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013273.hg.1 |
NM_021728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013273.hg.1 |
NM_172337.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013273.hg.1 |
TC14001167.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
ENST00000339475.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
ENST00000408990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
ENST00000554845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
ENST00000555006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
ENST00000555804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
NM_001270523.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
NM_001270524.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
NM_001270525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
NM_021728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
NM_172337.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
ENST00000339475.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
ENST00000408990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
ENST00000554845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
ENST00000555006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
ENST00000555804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
NM_001270523.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
NM_001270524.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
NM_001270525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
NM_021728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
NM_172337.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400168505.hg.1 |
TC1400009282.hg.1 |
Clariom D TC |