Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012329.hg.1 |
TC06001052.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012329.hg.1 |
ENST00000344281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012329.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012329.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012330.hg.1 |
TC06001052.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012330.hg.1 |
ENST00000344281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012330.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012330.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000406025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000406025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000585848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000585848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000585848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
ENST00000585848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000406025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000406025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000585848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000585848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000585848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
ENST00000585848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014186.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199508.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199508.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199508.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199508.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199508.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199508.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199508.hg.1 |
TC0600014186.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600199506.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199506.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199506.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199506.hg.1 |
ENST00000367591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199506.hg.1 |
ENST00000367592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199506.hg.1 |
NM_003630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199506.hg.1 |
TC0600014186.hg.1 |
Clariom D TC |