Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013915.hg.1 |
ENST00000328965.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013915.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013915.hg.1 |
TC11001098.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013917.hg.1 |
ENST00000328965.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013917.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013917.hg.1 |
TC11001098.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
ENST00000531220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
ENST00000531220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
ENST00000531220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
ENST00000531220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
ENST00000531220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009279.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013916.hg.1 |
ENST00000328965.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013916.hg.1 |
ENST00000531220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013916.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013916.hg.1 |
TC11001098.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100169509.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100169509.hg.1 |
ENST00000531220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100169509.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100169509.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100169509.hg.1 |
ENST00000531220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100169509.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100169509.hg.1 |
TC1100009279.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
ENST00000531220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
ENST00000531220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
ENST00000531220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
ENST00000531220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
ENST00000531220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009279.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100169510.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100169510.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100169510.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100169510.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100169510.hg.1 |
TC1100009279.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100169507.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100169507.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100169507.hg.1 |
ENST00000328965.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100169507.hg.1 |
NM_178507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100169507.hg.1 |
TC1100009279.hg.1 |
Clariom D TC |