Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008749.hg.1 |
TC0X000662.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008749.hg.1 |
ENST00000370634.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008749.hg.1 |
ENST00000430688.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008749.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008750.hg.1 |
TC0X000662.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008750.hg.1 |
ENST00000370634.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008750.hg.1 |
ENST00000430688.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008750.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008751.hg.1 |
TC0X000662.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008751.hg.1 |
ENST00000370634.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008751.hg.1 |
ENST00000430688.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008751.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158105.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158105.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158105.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158105.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158105.hg.1 |
TC0X00008534.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158107.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158107.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158107.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158107.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158107.hg.1 |
TC0X00008534.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00158104.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158104.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158104.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158104.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158104.hg.1 |
TC0X00008534.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00158106.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158106.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158106.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158106.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158106.hg.1 |
TC0X00008534.hg.1 |
Clariom D TC |