Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008284.hg.1 |
ENST00000354488.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008284.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008284.hg.1 |
ENST00000380213.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008284.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008284.hg.1 |
TC20000570.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008283.hg.1 |
ENST00000354488.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008283.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008283.hg.1 |
ENST00000380213.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008283.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008283.hg.1 |
TC20000570.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000470203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000470203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000470203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000470203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000470203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000496781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000496781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000496781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000496781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000496781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
ENST00000496781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008282.hg.1 |
ENST00000354488.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008282.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008282.hg.1 |
ENST00000380213.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008282.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008282.hg.1 |
TC20000570.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000157876.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157876.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157876.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157876.hg.1 |
TC2000008213.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000157876.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157876.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157876.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157877.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157877.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157877.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157877.hg.1 |
TC2000008213.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000157877.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157877.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157877.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157875.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157875.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157875.hg.1 |
ENST00000470203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157875.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157875.hg.1 |
TC2000008213.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000157875.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157875.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157875.hg.1 |
ENST00000470203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157875.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000354488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000380201.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000470203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000470203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000470203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000470203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000470203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000496781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000496781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000496781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000496781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000496781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
ENST00000496781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008213.hg.1 |
NM_023935.2 |
Refseq |