Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033701.hg.1 |
TC01002189.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033701.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033701.hg.1 |
ENST00000377223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033701.hg.1 |
ENST00000400903.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033701.hg.1 |
ENST00000488540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033701.hg.1 |
ENST00000541052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033701.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000471853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000471853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000471853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000471853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000488540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000488540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000488540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000488540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
ENST00000488540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012828.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033700.hg.1 |
TC01002189.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033700.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033700.hg.1 |
ENST00000377223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033700.hg.1 |
ENST00000400903.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033700.hg.1 |
ENST00000488540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033700.hg.1 |
ENST00000541052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033700.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033702.hg.1 |
TC01002189.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033702.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033702.hg.1 |
ENST00000377223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033702.hg.1 |
ENST00000400903.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033702.hg.1 |
ENST00000488540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033702.hg.1 |
ENST00000541052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033702.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000471853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000471853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000471853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000471853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000488540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000488540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000488540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000488540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
ENST00000488540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012828.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100194780.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194780.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194780.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194780.hg.1 |
ENST00000488540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194780.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100194780.hg.1 |
ENST00000377213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194780.hg.1 |
ENST00000377223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194780.hg.1 |
ENST00000400903.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194780.hg.1 |
ENST00000488540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194780.hg.1 |
NM_032368.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100194780.hg.1 |
TC0100012828.hg.1 |
Clariom D TC |