Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000468743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000468743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
ENST00000468743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016719.hg.1 |
TC09001334.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016719.hg.1 |
ENST00000262551.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016719.hg.1 |
ENST00000375561.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016719.hg.1 |
ENST00000468743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016719.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016719.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173488.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173488.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173488.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173488.hg.1 |
ENST00000468743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173488.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173488.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173488.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173488.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173488.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173488.hg.1 |
ENST00000468743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173488.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173488.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173488.hg.1 |
TC0900010794.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000468743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000468743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
ENST00000468743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010794.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173487.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173487.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173487.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173487.hg.1 |
ENST00000468743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173487.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173487.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173487.hg.1 |
ENST00000262551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173487.hg.1 |
ENST00000375561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173487.hg.1 |
ENST00000447356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173487.hg.1 |
ENST00000468743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173487.hg.1 |
NM_014057.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173487.hg.1 |
NM_033014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173487.hg.1 |
TC0900010794.hg.1 |
Clariom D TC |