Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015104.2 | NP_055919.2 | 22 - 23 | 3377 | 77 | |
XM_005273849.3 | XP_005273906.1 | 22 - 23 | 3344 | 77 | ||
XM_005273850.3 | XP_005273907.1 | 22 - 23 | 3344 | 77 | ||
XM_011544863.2 | XP_011543165.1 | 22 - 23 | 3368 | 77 | ||
XM_011544864.2 | XP_011543166.1 | 22 - 23 | 3353 | 77 | ||
XM_011544865.2 | XP_011543167.1 | 22 - 23 | 3361 | 77 | ||
XM_011544866.2 | XP_011543168.1 | 22 - 23 | 3368 | 77 | ||
XM_011544867.2 | XP_011543169.1 | 22 - 23 | 3368 | 77 | ||
XM_011544868.2 | XP_011543170.1 | 7 - 8 | 982 | 77 | ||
GenBank mRNA | AB007864.1 | - | 22 - 23 | 3318 | 77 | |
BC110650.1 | - | 22 - 23 | 3377 | 6147561 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |