Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC16A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276418.1 | NP_001263347.1 | 21 - 22 | 6470 | 106 | |
NM_014866.1 | NP_055681.1 | 22 - 23 | 6460 | 106 | ||
XM_005266134.2 | XP_005266191.1 | 21 - 22 | 6323 | 106 | ||
XM_011519246.2 | XP_011517548.1 | 22 - 23 | 6469 | 106 | ||
XM_011519247.1 | XP_011517549.1 | 21 - 22 | 6326 | 106 | ||
XM_011519248.2 | XP_011517550.1 | 21 - 22 | 6300 | 106 | ||
XM_011519249.1 | XP_011517551.1 | 21 - 22 | 6323 | 106 | ||
XM_011519250.2 | XP_011517552.1 | 21 - 22 | 6323 | 106 | ||
XM_011519251.1 | XP_011517553.1 | 21 - 22 | 6323 | 106 | ||
XM_011519252.2 | XP_011517554.1 | 21 - 22 | 6323 | 106 | ||
XM_011519253.1 | XP_011517555.1 | 21 - 22 | 6323 | 106 | ||
XM_011519254.1 | XP_011517556.1 | 21 - 22 | 6323 | 106 | ||
XM_011519255.2 | XP_011517557.1 | 21 - 22 | 6323 | 106 | ||
XM_011519256.1 | XP_011517558.1 | 21 - 22 | 6323 | 106 | ||
XM_011519257.1 | XP_011517559.1 | 21 - 22 | 6323 | 106 | ||
XM_011519258.1 | XP_011517560.1 | 20 - 21 | 6182 | 106 | ||
XM_011519259.1 | XP_011517561.1 | 21 - 22 | 6323 | 106 | ||
XM_011519260.2 | XP_011517562.1 | 21 - 22 | 6307 | 106 | ||
XM_011519261.1 | XP_011517563.1 | 18 - 19 | 5876 | 106 | ||
XM_011519262.1 | XP_011517564.1 | 22 - 23 | 5612 | 106 | ||
XM_011519263.2 | XP_011517565.1 | 22 - 23 | 5612 | 106 | ||
XM_017015363.1 | XP_016870852.1 | 21 - 22 | 6323 | 106 | ||
GenBank mRNA | AB002308.4 | - | 22 - 23 | 6460 | 106 | |
AB593115.1 | - | 21 - 22 | 6470 | 106 | ||
AK129634.1 | - | 1 - 2 | 73 | 106 | ||
BC001404.2 | - | 5 - 6 | 539 | 106 | ||
BC008332.1 | - | 11 - 12 | 1333 | 106 | ||
BC028183.1 | - | 17 - 18 | 2327 | 106 | ||
BC098454.1 | - | 5 - 6 | 623 | 106 | ||
BC125018.1 | - | 20 - 21 | 5032 | 106 | ||
BC125019.1 | - | 21 - 22 | 4321 | 106 | ||
DQ903855.1 | - | 20 - 21 | 5617 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |