Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023641.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023641.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023641.hg.1 |
TC10001790.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000184651.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184651.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000184651.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184651.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000184651.hg.1 |
TC1000012399.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
ENST00000368547.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012399.hg.1 |
NM_004092.3 |
Refseq |