Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs00931732_m1
See other SVIL GE Assays ›
Gene Symbol:
SVIL
Gene Name:
supervillin
Gene Aliases:
-
Chromosome Location:
Chr.10: 29457338 - 29737001 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs00931732_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: SVIL
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001323599.1
NP_001310528.1
14 - 15
2126
94
NM_001323600.1
NP_001310529.1
12 - 13
1727
94
NM_021738.2
NP_068506.2
13 - 14
3093
94
XM_005252570.2
XP_005252627.1
15 - 16
2990
94
XM_005252571.3
XP_005252628.1
15 - 16
2916
94
XM_005252573.2
XP_005252630.1
13 - 14
3122
94
XM_011519630.2
XP_011517932.2
13 - 14
3835
94
XM_011519632.2
XP_011517934.2
12 - 13
3664
94
XM_011519634.1
XP_011517936.1
15 - 16
3048
94
XM_011519635.1
XP_011517937.1
13 - 14
3015
94
XM_011519636.2
XP_011517938.2
12 - 13
2905
94
XM_011519637.2
XP_011517939.2
10 - 11
2653
94
XM_017016575.1
XP_016872064.1
13 - 14
3835
94
XM_017016576.1
XP_016872065.1
12 - 13
3754
94
XM_017016577.1
XP_016872066.1
13 - 14
3835
94
XM_017016579.1
XP_016872068.1
12 - 13
2905
94
XM_017016580.1
XP_016872069.1
11 - 12
2734
94
XM_017016581.1
XP_016872070.1
10 - 11
2653
94
XM_017016583.1
XP_016872072.1
13 - 14
3835
94
GenBank mRNA
AF109135.1
-
13 - 14
3093
94
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length between 86 and 100
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene SVIL
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
skeletal muscle tissue development
positive regulation of cytokinesis
protein binding
actin filament binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-zgjsk:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0