Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000325425.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000338555.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379101.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000379116.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000400928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000467651.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000467651.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000467651.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000470022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000470022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000470022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000470022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000470022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000470022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000470022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
ENST00000616713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NM_001130413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006497.hg.1 |
NR_037668.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144711.hg.1 |
TC0100006497.hg.1 |
Clariom D TC |