Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000506132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000506132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000506132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000506132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000506132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400145778.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145778.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145778.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145778.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400145778.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400145778.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145778.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145778.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145778.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400145778.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400145778.hg.1 |
TC0400006602.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000382774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000506132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000506132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000506132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000506132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000506132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000509689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
ENST00000511533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001297439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006602.hg.1 |
NM_001528.3 |
Refseq |