Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTH1P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | J04755.1 | - | 1 | 1071 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |