Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC2H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080463.1 | NP_001073932.1 | 72 - 73 | 10920 | 71 | |
NM_001377.2 | NP_001368.2 | 71 - 72 | 10899 | 71 | ||
XM_006718903.2 | XP_006718966.1 | 71 - 72 | 10878 | 71 | ||
XM_017018291.1 | XP_016873780.1 | 71 - 72 | 10899 | 71 | ||
XM_017018292.1 | XP_016873781.1 | 68 - 69 | 10153 | 71 | ||
GenBank mRNA | AB082528.2 | - | 23 - 24 | 2890 | 71 | |
AB290167.1 | - | 71 - 72 | 10953 | 71 | ||
AK095579.1 | - | 6 - 7 | 626 | 71 | ||
AK131453.1 | - | 16 - 17 | 1893 | 71 | ||
BC037496.1 | - | 8 - 9 | 891 | 71 | ||
BX538093.1 | - | 25 - 26 | 3030 | 71 | ||
BX649161.1 | - | 19 - 20 | 2274 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |