Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026759.hg.1 |
ENST00000260302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026759.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026759.hg.1 |
ENST00000546012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026759.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026759.hg.1 |
TC11002239.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000615555.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000615555.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000615555.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000615555.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000615555.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000615555.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000615555.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
ENST00000615555.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000615555.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000615555.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000615555.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000615555.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000615555.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000615555.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000615555.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
ENST00000615555.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012137.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193329.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193329.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193329.hg.1 |
ENST00000615555.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193329.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193329.hg.1 |
ENST00000260302.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193329.hg.1 |
ENST00000340273.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193329.hg.1 |
ENST00000615555.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193329.hg.1 |
NM_002427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193329.hg.1 |
TC1100012137.hg.1 |
Clariom D TC |