Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000485253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000485253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000485253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000485253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000485253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000485253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000485253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000485253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000495403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000495403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000495403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
ENST00000495403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300157689.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157689.hg.1 |
ENST00000485253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157689.hg.1 |
ENST00000495403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157689.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300157689.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157689.hg.1 |
ENST00000485253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157689.hg.1 |
ENST00000495403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157689.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300157689.hg.1 |
TC0300008088.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000389622.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000485253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000485253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000485253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000485253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000485253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000485253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000485253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000485253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000495403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000495403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000495403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
ENST00000495403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008088.hg.1 |
NM_153605.3 |
Refseq |