Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025738.hg.1 |
TC03001429.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025738.hg.1 |
ENST00000359365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025738.hg.1 |
NM_001206957.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025738.hg.1 |
NM_007182.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025738.hg.1 |
NM_170714.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025736.hg.1 |
TC03001429.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025736.hg.1 |
ENST00000357043.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025736.hg.1 |
ENST00000359365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025736.hg.1 |
ENST00000488024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025736.hg.1 |
NM_001206957.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025736.hg.1 |
NM_007182.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025736.hg.1 |
NM_170714.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182697.hg.1 |
ENST00000359365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182697.hg.1 |
ENST00000478619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182697.hg.1 |
ENST00000482447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182697.hg.1 |
ENST00000616212.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182697.hg.1 |
NM_001206957.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182697.hg.1 |
NM_007182.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182697.hg.1 |
NM_170714.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182697.hg.1 |
ENST00000359365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182697.hg.1 |
ENST00000478619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182697.hg.1 |
ENST00000482447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182697.hg.1 |
ENST00000616212.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182697.hg.1 |
NM_001206957.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182697.hg.1 |
NM_007182.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182697.hg.1 |
NM_170714.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182697.hg.1 |
TC0300011105.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
ENST00000357043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
ENST00000359365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
ENST00000395117.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
ENST00000478619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
ENST00000482447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
ENST00000488024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
ENST00000616212.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
NM_001206957.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
NM_007182.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
NM_170714.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
ENST00000357043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
ENST00000359365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
ENST00000395117.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
ENST00000478619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
ENST00000482447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
ENST00000488024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
ENST00000616212.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
NM_001206957.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
NM_007182.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
NM_170714.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182694.hg.1 |
TC0300011105.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
ENST00000357043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
ENST00000359365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
ENST00000395117.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
ENST00000478619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
ENST00000482447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
ENST00000488024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
ENST00000616212.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
NM_001206957.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
NM_007182.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
NM_170714.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
ENST00000357043.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
ENST00000359365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
ENST00000395117.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
ENST00000478619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
ENST00000482447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
ENST00000488024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
ENST00000616212.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
NM_001206957.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
NM_007182.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
NM_170714.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182695.hg.1 |
TC0300011105.hg.1 |
Clariom D TC |