Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs00948652_m1
See other PPFIBP1 GE Assays ›
Gene Symbol:
PPFIBP1
Gene Name:
PPFIA binding protein 1
Gene Aliases:
L2, SGT2, hSGT2, hSgt2p
Chromosome Location:
Chr.12: 27524112 - 27695564 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs00948652_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: PPFIBP1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_003622.3
NP_003613.3
12 - 13
1371
64
XM_005253505.1
XP_005253562.1
12 - 13
1167
64
XM_005253508.1
XP_005253565.1
12 - 13
1167
64
XM_005253509.1
XP_005253566.1
11 - 12
1134
64
XM_006719159.3
XP_006719222.1
11 - 12
1121
64
XM_017020056.1
XP_016875545.1
14 - 15
1483
64
XM_017020057.1
XP_016875546.1
12 - 13
1658
64
XM_017020059.1
XP_016875548.1
13 - 14
1450
64
XM_017020060.1
XP_016875549.1
12 - 13
1167
64
XM_017020062.1
XP_016875551.1
12 - 13
1414
64
XM_017020063.1
XP_016875552.1
12 - 13
1417
64
XM_017020066.1
XP_016875555.1
12 - 13
1340
64
XM_017020067.1
XP_016875556.1
11 - 12
1307
64
XM_017020068.1
XP_016875557.1
12 - 13
1356
64
XM_017020071.1
XP_016875560.1
12 - 13
1340
64
XM_017020072.1
XP_016875561.1
11 - 12
1307
64
XM_017020074.1
XP_016875563.1
10 - 11
1108
64
XM_017020075.1
XP_016875564.1
10 - 11
1006
64
XM_017020076.1
XP_016875565.1
2 - 3
149
64
XM_017020077.1
XP_016875566.1
12 - 13
1167
64
XM_017020078.1
XP_016875567.1
12 - 13
1345
64
GenBank mRNA
AF034802.1
-
12 - 13
1234
64
AK294267.1
-
9 - 10
951
64
Back To Top
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact
Tech Support
if you need more information.
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene PPFIBP1
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
cell-cell adhesion
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0