Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003258.hg.1 |
ENST00000228425.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003258.hg.1 |
ENST00000318304.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003258.hg.1 |
ENST00000537927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003258.hg.1 |
ENST00000542629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003258.hg.1 |
ENST00000543820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003258.hg.1 |
NM_001198915.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003258.hg.1 |
NM_001198916.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003258.hg.1 |
NM_003622.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003258.hg.1 |
NM_177444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003258.hg.1 |
TC12000266.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
ENST00000228425.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
ENST00000318304.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
ENST00000537927.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
ENST00000540114.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
ENST00000542629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
NM_001198915.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
NM_001198916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
NM_003622.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
NM_177444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
ENST00000228425.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
ENST00000318304.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
ENST00000537927.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
ENST00000540114.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
ENST00000542629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
NM_001198915.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
NM_001198916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
NM_003622.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
NM_177444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200150239.hg.1 |
TC1200007153.hg.1 |
Clariom D TC |