Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011884.hg.1 |
TC22001489.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011884.hg.1 |
ENST00000266095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011884.hg.1 |
ENST00000336566.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011884.hg.1 |
ENST00000382151.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011884.hg.1 |
ENST00000397500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011884.hg.1 |
ENST00000439502.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011884.hg.1 |
ENST00000451635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011884.hg.1 |
ENST00000478893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011884.hg.1 |
NM_014338.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
TC2200008487.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000266095.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000382151.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000397500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000422020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000435900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000437808.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000439502.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000460723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000473770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000474017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000478893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000491342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
NM_014338.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000266095.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000382151.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000397500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000422020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000435900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000437808.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000439502.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000460723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000473770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000474017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000478893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
ENST00000491342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161099.hg.1 |
NM_014338.3 |
Refseq |