Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018140.hg.1 |
TC03001024.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018140.hg.1 |
ENST00000265023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018140.hg.1 |
ENST00000287611.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018140.hg.1 |
ENST00000432028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018140.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018140.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171132.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171132.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171132.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171132.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171132.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171132.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171132.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171132.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171132.hg.1 |
TC0300009777.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000265023.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000287611.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
ENST00000447445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_000893.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001102416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009777.hg.1 |
NM_001166451.1 |
Refseq |