Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022037.hg.1 |
TC06001825.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022037.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022037.hg.1 |
ENST00000446683.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022037.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012113.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183388.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183388.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183388.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183388.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183388.hg.1 |
TC0600012113.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
ENST00000370830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
NM_021073.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012113.hg.1 |
NM_021073.3 |
Refseq |