Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000463225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000463225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000463225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000463225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000463225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000463225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000483367.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000483367.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000483367.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000483367.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000483367.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
ENST00000483367.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026305.hg.1 |
TC06004137.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026305.hg.1 |
ENST00000275189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026305.hg.1 |
ENST00000368149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026305.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600190715.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600190715.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600190715.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600190715.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600190715.hg.1 |
TC0600013147.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000368149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000463225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000463225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000463225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000463225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000463225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000463225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000483367.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000483367.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000483367.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000483367.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000483367.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
ENST00000483367.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013147.hg.1 |
NM_033515.2 |
Refseq |