Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011174.hg.1 |
TC08000843.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011174.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011174.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000628266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000628266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800163178.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163178.hg.1 |
ENST00000525101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163178.hg.1 |
ENST00000530409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163178.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163178.hg.1 |
ENST00000534255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163178.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800163178.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163178.hg.1 |
ENST00000525101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163178.hg.1 |
ENST00000530409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163178.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163178.hg.1 |
ENST00000534255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163178.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800163178.hg.1 |
TC0800009242.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000628266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000628266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |