Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000206.hg.1 |
TC01000022.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000206.hg.1 |
ENST00000379407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000206.hg.1 |
ENST00000379409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000206.hg.1 |
ENST00000379410.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000206.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000206.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000480267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000480267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000480267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000480267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000480267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000491024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000491024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000491024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000491024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
ENST00000491024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144422.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144422.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144422.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144422.hg.1 |
ENST00000480267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144422.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144422.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144422.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144422.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144422.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144422.hg.1 |
ENST00000480267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144422.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144422.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144422.hg.1 |
TC0100006480.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379407.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379409.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000379410.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000480267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000480267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000480267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000480267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000480267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000491024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000491024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000491024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000491024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
ENST00000491024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_001160184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006480.hg.1 |
NM_032129.2 |
Refseq |