Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000494752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000494752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000494752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000494752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000494752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000494752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000494752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
ENST00000494752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012964.hg.1 |
ENST00000371582.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012964.hg.1 |
ENST00000371583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012964.hg.1 |
ENST00000371588.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012964.hg.1 |
ENST00000449701.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012964.hg.1 |
ENST00000466152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012964.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012964.hg.1 |
TC20000940.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012963.hg.1 |
ENST00000371582.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012963.hg.1 |
ENST00000371583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012963.hg.1 |
ENST00000371588.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012963.hg.1 |
ENST00000449701.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012963.hg.1 |
ENST00000466152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012963.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012963.hg.1 |
TC20000940.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012965.hg.1 |
ENST00000371582.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012965.hg.1 |
ENST00000371583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012965.hg.1 |
ENST00000371588.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012965.hg.1 |
ENST00000449701.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012965.hg.1 |
ENST00000466152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012965.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012965.hg.1 |
TC20000940.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000166072.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166072.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166072.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166072.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166072.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166072.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000166072.hg.1 |
TC2000009447.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000166072.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166072.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166072.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166072.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166072.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166072.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000166071.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166071.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166071.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166071.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166071.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166071.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000166071.hg.1 |
TC2000009447.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000166071.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166071.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166071.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166071.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166071.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166071.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371582.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000371588.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000413082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000466152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000494752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000494752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000494752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000494752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000494752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000494752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000494752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
ENST00000494752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009447.hg.1 |
NM_003859.2 |
Refseq |