Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009450.hg.1 |
TC07003314.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009450.hg.1 |
ENST00000198536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009450.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009450.hg.1 |
ENST00000394000.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009450.hg.1 |
ENST00000453419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009450.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009450.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009450.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000394000.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000394000.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000394000.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000394000.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000394000.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000394000.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000394000.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000474013.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000474013.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000484934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000484934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000484934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
ENST00000484934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013429.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000394000.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000394000.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000394000.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000394000.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000394000.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000394000.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000394000.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000474013.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000474013.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000484934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000484934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000484934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
ENST00000484934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013429.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198378.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198378.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198378.hg.1 |
ENST00000394000.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198378.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198378.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198378.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198378.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198378.hg.1 |
ENST00000198536.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198378.hg.1 |
ENST00000350573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198378.hg.1 |
ENST00000394000.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198378.hg.1 |
ENST00000453419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198378.hg.1 |
NM_013439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198378.hg.1 |
NM_178272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198378.hg.1 |
NM_178273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198378.hg.1 |
TC0700013429.hg.1 |
Clariom D TC |