Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014241.hg.1 |
ENST00000227349.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014241.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014241.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014241.hg.1 |
TC11001117.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014243.hg.1 |
ENST00000227349.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014243.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014243.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014243.hg.1 |
TC11001117.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014242.hg.1 |
ENST00000227349.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014242.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014242.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014242.hg.1 |
TC11001117.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000534362.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000534362.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000534362.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170085.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170085.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170085.hg.1 |
ENST00000534362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170085.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170085.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170085.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170085.hg.1 |
ENST00000534362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170085.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170085.hg.1 |
TC1100009335.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100170087.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170087.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170087.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170087.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170087.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170087.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170087.hg.1 |
TC1100009335.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000534362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000534362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000534362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170084.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170084.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170084.hg.1 |
ENST00000534362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170084.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170084.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170084.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170084.hg.1 |
ENST00000534362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170084.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170084.hg.1 |
TC1100009335.hg.1 |
Clariom D TC |