Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033274.4 | NP_150377.1 | 17 - 18 | 2117 | 97 | |
XM_005266003.2 | XP_005266060.1 | 17 - 18 | 2069 | 97 | ||
XM_011534682.2 | XP_011532984.1 | 10 - 11 | 2873 | 97 | ||
XM_017010009.1 | XP_016865498.1 | 10 - 11 | 1334 | 97 | ||
GenBank mRNA | AF134707.1 | - | 14 - 15 | 1698 | 97 | |
AF311317.1 | - | 17 - 18 | 2026 | 97 | ||
AF326918.1 | - | 17 - 18 | 2038 | 97 | ||
AK091464.1 | - | 17 - 18 | 2079 | 97 | ||
AK308182.1 | - | 17 - 18 | 2069 | 97 | ||
BC024214.2 | - | 6 - 7 | 702 | 97 | ||
BC033132.1 | - | 6 - 7 | 702 | 97 | ||
Y13786.2 | - | 17 - 18 | 2067 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |