Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000443638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000443638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000443638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000443638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000443638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000443638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000443638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000443638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000464654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000464654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000464654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000464654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000464654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000464654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000475401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000475401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000475401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000485093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000485093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000485093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000485093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000485093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000490192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000490192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000490192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000490192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
ENST00000490192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019776.hg.1 |
ENST00000371142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019776.hg.1 |
ENST00000490192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019776.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019776.hg.1 |
TC10001558.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000178334.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178334.hg.1 |
ENST00000443638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178334.hg.1 |
ENST00000490192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178334.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178334.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178334.hg.1 |
ENST00000443638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178334.hg.1 |
ENST00000490192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178334.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178334.hg.1 |
TC1000011522.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000371142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000443638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000443638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000443638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000443638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000443638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000443638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000443638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000443638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000464654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000464654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000464654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000464654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000464654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000464654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000475401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000475401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000475401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000485093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000485093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000485093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000485093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000485093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000490192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000490192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000490192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000490192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
ENST00000490192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011522.hg.1 |
NM_020123.3 |
Refseq |