Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15011848.hg.1 |
ENST00000452443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15011848.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15011848.hg.1 |
TC15001275.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
ENST00000562372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
ENST00000562372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
ENST00000562372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500167146.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500167146.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500167146.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500167146.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500167146.hg.1 |
TC1500009216.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
ENST00000452443.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
ENST00000562372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
ENST00000562372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
ENST00000562372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009216.hg.1 |
NM_052955.2 |
Refseq |