Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081955.2 | NP_001075424.1 | 9 - 10 | 845 | 85 | |
NM_001165933.1 | NP_001159405.1 | 9 - 10 | 845 | 85 | ||
GenBank mRNA | AK290535.1 | - | 9 - 10 | 714 | 85 | |
AK309061.1 | - | 9 - 10 | 735 | 85 | ||
AY585191.1 | - | 9 - 10 | 642 | 85 | ||
BC022504.1 | - | 9 - 10 | 712 | 4801328 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |