Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010401.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010401.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010401.hg.1 |
TC20000779.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010400.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010400.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010400.hg.1 |
TC20000779.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008909.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162087.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162087.hg.1 |
NM_003908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162087.hg.1 |
TC2000008909.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000162087.hg.1 |
ENST00000374980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162087.hg.1 |
NM_003908.4 |
Refseq |