Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YTHDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017009733.1 | XP_016865222.1 | 2 - 3 | 488 | 138 | |
XM_017009734.1 | XP_016865223.1 | 2 - 3 | 488 | 138 | ||
XM_017009735.1 | XP_016865224.1 | 2 - 3 | 456 | 138 | ||
GenBank mRNA | AK096486.1 | - | 2 - 3 | 477 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |