Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000527558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000527558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000527558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000527558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000527558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028245.hg.1 |
ENST00000264029.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028245.hg.1 |
ENST00000397925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028245.hg.1 |
ENST00000450700.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028245.hg.1 |
ENST00000525958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028245.hg.1 |
ENST00000529101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028245.hg.1 |
ENST00000530256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028245.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028245.hg.1 |
TC11002352.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100196096.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100196096.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100196096.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100196096.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100196096.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100196096.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100196096.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100196096.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100196096.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100196096.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100196096.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100196096.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100196096.hg.1 |
TC1100012492.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000264029.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000397925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000527558.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000527558.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000527558.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000527558.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000527558.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000531295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
ENST00000613915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_001301065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012492.hg.1 |
NM_007180.2 |
Refseq |