Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003480.3 | NP_003471.1 | 6 - 7 | 477 | 87 | |
NR_123733.1 | 6 - 7 | 477 | 87 | |||
GenBank mRNA | AK290585.1 | - | 6 - 7 | 268 | 87 | |
AK315807.1 | - | 6 - 7 | 264 | 87 | ||
AU280458.1 | - | 6 - 7 | 415 | 87 | ||
BC005901.1 | - | 6 - 7 | 267 | 4247343 | 87 | |
DN996275.1 | - | 6 - 7 | 298 | 87 | ||
U37283.1 | - | 6 - 7 | 399 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |