Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007696.hg.1 |
ENST00000361840.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007696.hg.1 |
ENST00000492258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007696.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007696.hg.1 |
TC13000667.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007695.hg.1 |
ENST00000443861.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007695.hg.1 |
ENST00000443861.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000492258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000492258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000613924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000613924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000613924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000613924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000613924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000613924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
ENST00000613924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NR_023351.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NR_023351.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NR_023351.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NR_023351.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NR_023351.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NR_023351.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NR_023351.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NR_023351.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008936.hg.1 |
NR_023351.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300160305.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300160305.hg.1 |
ENST00000492258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300160305.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300160305.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300160305.hg.1 |
ENST00000492258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300160305.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300160305.hg.1 |
TC1300008936.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00007695.hg.1 |
ENST00000443861.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007695.hg.1 |
ENST00000443861.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000361840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000378195.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000492258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000492258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000613924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000613924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000613924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000613924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000613924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000613924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
ENST00000613924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_001127482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NM_020456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NR_023351.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NR_023351.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NR_023351.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NR_023351.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NR_023351.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NR_023351.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NR_023351.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NR_023351.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008936.hg.1 |
NR_023351.2 |
Refseq |