Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052581.hg.1 |
TC01003363.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052581.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052581.hg.1 |
ENST00000448509.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052581.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221841.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221841.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221841.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221841.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221841.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221841.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221841.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221841.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221841.hg.1 |
TC0100016045.hg.1 |
Clariom D TC |