Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01051021.hg.1 |
TC01003301.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01051021.hg.1 |
ENST00000295536.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01051021.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01051021.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000467991.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000467991.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000467991.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000467991.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000467991.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000498036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000498036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000498036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000498036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000498036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
ENST00000498036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000467991.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000467991.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000467991.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000467991.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000467991.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000498036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000498036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000498036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000498036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000498036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
ENST00000498036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015939.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100220075.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100220075.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100220075.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100220075.hg.1 |
ENST00000368424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100220075.hg.1 |
ENST00000485982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100220075.hg.1 |
NM_144622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100220075.hg.1 |
TC0100015939.hg.1 |
Clariom D TC |