Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
ENST00000598738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
ENST00000598738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
ENST00000598738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
ENST00000598738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
ENST00000598738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003821.hg.1 |
ENST00000221740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003821.hg.1 |
ENST00000427043.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003821.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003821.hg.1 |
TC19000266.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900152046.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152046.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900152046.hg.1 |
TC1900007205.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900152046.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152046.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
ENST00000427043.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
ENST00000598738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
ENST00000598738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
ENST00000598738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
ENST00000598738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
ENST00000598738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007205.hg.1 |
NM_012114.2 |
Refseq |